Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198309.1 | NP_001185238.1 | 13 - 14 | 3134 | 68 | |
NM_001333706.1 | NP_001319240.1 | 13 - 14 | 3149 | 68 | ||
NM_001333707.1 | NP_001321273.1 | 13 - 14 | 3146 | 68 | ||
NM_104411.4 | NP_175932.2 | 13 - 14 | 3146 | 68 | ||
NM_180635.1 | NP_850966.1 | 12 - 13 | 2823 | 68 | ||
NM_180636.1 | NP_850967.1 | 12 - 13 | 2823 | 68 | ||
GenBank mRNA | AY061803.1 | - | 12 - 13 | 2821 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |