Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120500.3 | NP_196038.1 | 5 - 6 | 762 | 73 | |
GenBank mRNA | AK221569.1 | - | 5 - 6 | 762 | 73 | |
AY088872.1 | - | 5 - 6 | 758 | 73 | ||
DQ446916.1 | - | 5 - 6 | 715 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |