Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342767.1 | NP_001330089.1 | 10 - 11 | 1407 | 102 | |
NM_120518.5 | NP_196056.2 | 10 - 11 | 1420 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |