Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120572.3 | NP_568145.1 | 9 - 10 | 846 | 86 | |
GenBank mRNA | AB255027.1 | - | 9 - 10 | 794 | 86 | |
AY085229.1 | - | 9 - 10 | 805 | 86 | ||
BT025238.1 | - | 9 - 10 | 794 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |