Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G18E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120597.2 | NP_196134.1 | 2 - 3 | 1016 | 64 | |
GenBank mRNA | DQ056670.1 | - | 2 - 3 | 1016 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |