Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125703.2 | NP_001119175.1 | 10 - 11 | 1383 | 80 | |
NM_120636.4 | NP_196173.1 | 11 - 12 | 1297 | 80 | ||
GenBank mRNA | AK118437.1 | - | 11 - 12 | 1273 | 80 | |
BT009685.1 | - | 11 - 12 | 1160 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |