Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036763.2 | NP_001031840.1 | 13 - 14 | 1495 | 105 | |
NM_120680.3 | NP_196217.2 | 14 - 15 | 1437 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |