Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AtPOT1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120714.4 | NP_196249.2 | 3 - 4 | 577 | 145 | |
GenBank mRNA | AK221379.1 | - | 3 - 4 | 529 | 145 | |
AY884594.1 | - | 3 - 4 | 452 | 145 | ||
BT012568.1 | - | 3 - 4 | 452 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |