Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WCRKC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036767.2 | NP_001031844.1 | 1 - 2 | 499 | 94 | |
NM_001342898.1 | NP_001330197.1 | 1 - 2 | 445 | 94 | ||
NM_001342899.1 | NP_001330198.1 | 1 - 2 | 499 | 94 | ||
NM_120752.4 | NP_568172.1 | 1 - 2 | 499 | 94 | ||
GenBank mRNA | BT029447.1 | - | 1 - 2 | 374 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |