Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCBK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343803.1 | NP_001331260.1 | 5 - 6 | 1291 | 113 | |
NM_001343804.1 | NP_001331261.1 | 6 - 7 | 1273 | 113 | ||
NM_122252.4 | NP_568432.1 | 5 - 6 | 1445 | 113 | ||
NM_180734.3 | NP_851065.1 | 5 - 6 | 1311 | 113 | ||
GenBank mRNA | AB046717.1 | - | 4 - 5 | 951 | 113 | |
AY139759.1 | - | 5 - 6 | 1367 | 113 | ||
BT004544.1 | - | 4 - 5 | 951 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |