Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOPP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036923.1 | NP_001032000.1 | 1 - 2 | 327 | 84 | |
NM_123704.3 | NP_568625.1 | 1 - 2 | 327 | 84 | ||
NM_180792.3 | NP_851123.1 | 1 - 2 | 327 | 84 | ||
GenBank mRNA | AY136464.1 | - | 1 - 2 | 327 | 84 | |
BT002143.1 | - | 1 - 2 | 171 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |