Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344577.1 | NP_001331586.1 | 1 - 2 | 779 | 93 | |
NM_123861.3 | NP_199306.1 | 1 - 2 | 791 | 93 | ||
NM_180801.2 | NP_851132.1 | 1 - 2 | 791 | 93 | ||
GenBank mRNA | AY048259.1 | - | 1 - 2 | 779 | 93 | |
BT002634.1 | - | 1 - 2 | 776 | 93 | ||
KJ138648.1 | - | 1 - 2 | 776 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |