Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036960.2 | NP_001032037.1 | 12 - 13 | 1154 | 93 | |
NM_001344808.1 | NP_001331840.1 | 12 - 13 | 1326 | 93 | ||
NM_124265.5 | NP_568704.2 | 11 - 12 | 2021 | 93 | ||
NM_180826.4 | NP_851157.1 | 11 - 12 | 1171 | 93 | ||
GenBank mRNA | AF062590.1 | - | 11 - 12 | 1051 | 93 | |
AF062591.1 | - | 11 - 12 | 1270 | 93 | ||
AY079026.1 | - | 11 - 12 | 1156 | 93 | ||
AY136455.1 | - | 11 - 12 | 1059 | 93 | ||
BT002594.1 | - | 10 - 11 | 958 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |