Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHOT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345288.1 | NP_001318824.1 | 14 - 15 | 2423 | 54 | |
NM_001345289.1 | NP_001330355.1 | 13 - 14 | 2062 | 54 | ||
NM_001345290.1 | NP_001330356.1 | 12 - 13 | 2412 | 54 | ||
NM_125199.2 | NP_568874.2 | 13 - 14 | 1882 | 54 | ||
NM_180879.2 | NP_851210.1 | 13 - 14 | 2239 | 54 | ||
NM_180880.2 | NP_851211.1 | 13 - 14 | 2235 | 54 | ||
NM_180881.3 | NP_851212.2 | 12 - 13 | 2020 | 54 | ||
GenBank mRNA | AF053941.2 | - | 12 - 13 | 1777 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |