Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZEU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085303.1 | NP_001078772.1 | 4 - 5 | 866 | 65 | |
NM_125335.4 | NP_568907.2 | 4 - 5 | 987 | 65 | ||
NM_180891.4 | NP_851222.1 | 4 - 5 | 840 | 65 | ||
GenBank mRNA | AK226501.1 | - | 4 - 5 | 802 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |