Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125995.2 | NP_001119467.1 | 14 - 15 | 1838 | 61 | |
NM_001345427.1 | NP_001332136.1 | 14 - 15 | 2029 | 61 | ||
NM_001345428.1 | NP_001332137.1 | 14 - 15 | 1925 | 61 | ||
NM_125453.6 | NP_200868.2 | 14 - 15 | 1754 | 61 | ||
NM_180902.5 | NP_851233.1 | 14 - 15 | 1838 | 61 | ||
GenBank mRNA | AF434673.1 | - | 14 - 15 | 1694 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |