Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345460.1 | NP_001330163.1 | 7 - 8 | 1236 | 84 | |
NM_125518.5 | NP_200933.2 | 8 - 9 | 1295 | 84 | ||
NM_180907.3 | NP_851238.1 | 8 - 9 | 1297 | 84 | ||
GenBank mRNA | AK317449.1 | - | 8 - 9 | 1287 | 84 | |
AY099736.1 | - | 8 - 9 | 1218 | 84 | ||
BT002603.1 | - | 7 - 8 | 1137 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |