Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125784.3 | NP_568981.2 | 5 - 6 | 512 | 77 | |
NM_180929.4 | NP_851260.1 | 4 - 5 | 539 | 77 | ||
GenBank mRNA | AB027709.1 | - | 5 - 6 | 459 | 77 | |
AY039881.1 | - | 5 - 6 | 501 | 77 | ||
AY143900.1 | - | 5 - 6 | 410 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |