Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATH13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180940.2 | NP_851271.1 | 19 - 20 | 2459 | 136 | |
GenBank mRNA | AY059877.1 | - | 19 - 20 | 2442 | 136 | |
AY064044.1 | - | 19 - 20 | 2418 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |