Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZWI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203699.1 | NP_001190628.1 | 20 - 21 | 3680 | 82 | |
NM_125990.4 | NP_569022.2 | 20 - 21 | 3787 | 82 | ||
NM_180945.2 | NP_851276.1 | 20 - 21 | 3732 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |