Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035765.2 | NP_001030842.1 | 8 - 9 | 865 | 80 | |
NM_001339563.1 | NP_001326925.1 | 8 - 9 | 829 | 80 | ||
NM_001339564.1 | NP_001326924.1 | 5 - 6 | 462 | 80 | ||
NM_115078.4 | NP_566960.1 | 8 - 9 | 961 | 80 | ||
GenBank mRNA | AJ302781.1 | - | 8 - 9 | 746 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |