Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OEP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115102.3 | NP_190810.1 | 1 | 226 | 58 | |
GenBank mRNA | AK175275.1 | - | 1 | 168 | 58 | |
AK175812.1 | - | 1 | 180 | 58 | ||
BT010511.1 | - | 1 | 137 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |