Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWINV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035770.2 | NP_001030847.1 | 3 - 4 | 1704 | 68 | |
NM_001339580.1 | NP_001325488.1 | 3 - 4 | 1871 | 68 | ||
NM_115120.4 | NP_190828.2 | 4 - 5 | 1672 | 68 | ||
GenBank mRNA | DQ446760.1 | - | 4 - 5 | 1585 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |