Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203156.1 | NP_001190085.1 | 22 - 23 | 5149 | 73 | |
NM_115288.8 | NP_190996.3 | 21 - 22 | 5180 | 73 | ||
GenBank mRNA | AB440793.1 | - | 20 - 21 | 4918 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |