Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035780.3 | NP_001030857.1 | 5 - 6 | 844 | 87 | |
NM_001339658.1 | NP_001319744.1 | 3 - 4 | 260 | 87 | ||
NM_001339659.1 | NP_001325502.1 | 4 - 5 | 973 | 87 | ||
NM_001339660.1 | NP_001325503.1 | 3 - 4 | 355 | 87 | ||
NM_115292.4 | NP_191000.3 | 5 - 6 | 648 | 87 | ||
GenBank mRNA | AY254038.2 | - | 5 - 6 | 602 | 87 | |
AY885245.1 | - | 5 - 6 | 523 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |