Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPB5E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115306.5 | NP_191013.1 | 1 - 2 | 356 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Arabidopsis thaliana Probe spans exons |