Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115370.4 | NP_191072.1 | 2 - 3 | 476 | 74 | |
GenBank mRNA | AJ418046.1 | - | 2 - 3 | 345 | 74 | |
BT004265.1 | - | 2 - 3 | 345 | 74 | ||
BT005528.1 | - | 2 - 3 | 294 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |