Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115391.5 | NP_191093.1 | 4 - 5 | 541 | 122 | |
GenBank mRNA | AF419553.1 | - | 4 - 5 | 500 | 122 | |
AY087663.1 | - | 4 - 5 | 436 | 122 | ||
AY097338.1 | - | 4 - 5 | 424 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |