Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P5CS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125374.1 | NP_001118846.1 | 13 - 14 | 1637 | 81 | |
NM_115419.5 | NP_191120.2 | 13 - 14 | 1839 | 81 | ||
GenBank mRNA | AY091766.1 | - | 13 - 14 | 1637 | 81 | |
Y09355.1 | - | 1607 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |