Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115625.4 | NP_567052.1 | 7 - 8 | 976 | 81 | |
GenBank mRNA | BT026036.1 | - | 7 - 8 | 608 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |