Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344487.1 | NP_001331886.1 | 12 - 13 | 1676 | 132 | |
NM_001344488.1 | NP_001331887.1 | 12 - 13 | 1518 | 132 | ||
NM_123716.3 | NP_199163.1 | 12 - 13 | 1630 | 132 | ||
NM_203147.2 | NP_974876.1 | 12 - 13 | 1676 | 132 | ||
GenBank mRNA | AY057568.1 | - | 12 - 13 | 1445 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |