Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUVR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085233.2 | NP_001078702.1 | 7 - 8 | 2077 | 87 | |
NM_001085234.2 | NP_001078703.1 | 7 - 8 | 2164 | 87 | ||
NM_001203540.1 | NP_001190469.1 | 7 - 8 | 2200 | 87 | ||
NM_001344513.1 | NP_001331768.1 | 7 - 8 | 2178 | 87 | ||
NM_001344514.1 | NP_001331767.1 | 7 - 8 | 2178 | 87 | ||
NM_001344515.1 | NP_001331773.1 | 6 - 7 | 2236 | 87 | ||
NM_001344516.1 | NP_001331766.1 | 6 - 7 | 2162 | 87 | ||
NM_001344517.1 | NP_001331772.1 | 7 - 8 | 2102 | 87 | ||
NM_001344518.1 | NP_001331771.1 | 7 - 8 | 2088 | 87 | ||
NM_001344519.1 | NP_001331770.1 | 7 - 8 | 2088 | 87 | ||
NM_001344520.1 | NP_001331769.1 | 7 - 8 | 2014 | 87 | ||
NM_001344521.1 | NP_001331765.1 | 6 - 7 | 2326 | 87 | ||
NM_001344522.1 | NP_001331764.1 | 6 - 7 | 2266 | 87 | ||
NM_001344523.1 | NP_001331763.1 | 6 - 7 | 2252 | 87 | ||
NM_001344524.1 | NP_001331762.1 | 7 - 8 | 2238 | 87 | ||
NM_123766.3 | NP_568631.1 | 7 - 8 | 2090 | 87 | ||
NM_203151.3 | NP_974880.1 | 7 - 8 | 2131 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |