Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103365.1 | NP_174902.1 | 1 - 2 | 290 | 143 | |
GenBank mRNA | DQ056481.1 | - | 1 - 2 | 290 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |