Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123962.2 | NP_001117434.1 | 8 - 9 | 2116 | 132 | |
NM_103511.4 | NP_175051.1 | 8 - 9 | 2377 | 132 | ||
GenBank mRNA | AF378782.1 | - | 8 - 9 | 2213 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |