Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339909.1 | NP_001319790.1 | 24 - 25 | 3766 | 122 | |
NM_001339910.1 | NP_001327763.1 | 24 - 25 | 3927 | 122 | ||
NM_115678.2 | NP_191375.1 | 23 - 24 | 3415 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |