Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203206.1 | NP_001190135.1 | 21 - 22 | 3319 | 59 | |
NM_001339969.1 | NP_001326041.1 | 21 - 22 | 4186 | 59 | ||
NM_001339970.1 | NP_001326042.1 | 20 - 21 | 3328 | 59 | ||
NM_001339971.1 | NP_001326040.1 | 20 - 21 | 3099 | 59 | ||
NM_115803.3 | NP_191500.2 | 20 - 21 | 3218 | 59 | ||
GenBank mRNA | AJ459823.1 | - | 20 - 21 | 2866 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |