Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSY5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115873.3 | NP_191569.2 | 7 - 8 | 615 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |