Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKKK14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128559.3 | NP_180565.1 | 1 | 1313 | 74 | |
GenBank mRNA | BT000903.1 | - | 1 | 1135 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |