Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336238.1 | NP_001324058.1 | 5 - 6 | 1396 | 88 | |
NM_128562.4 | NP_180568.1 | 5 - 6 | 1091 | 88 | ||
GenBank mRNA | AF029876.1 | - | 5 - 6 | 862 | 88 | |
AF033118.1 | - | 5 - 6 | 896 | 88 | ||
AK221387.1 | - | 5 - 6 | 958 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |