Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121376.4 | NP_196877.1 | 1 - 2 | 567 | 103 | |
GenBank mRNA | AF101075.1 | - | 1 - 2 | 500 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |