Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARAB-AK-LYS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085112.2 | NP_001078581.1 | 7 - 8 | 1340 | 68 | |
NM_001343314.1 | NP_001330615.1 | 7 - 8 | 1309 | 68 | ||
NM_121409.4 | NP_196910.1 | 7 - 8 | 1343 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |