Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121491.3 | NP_196991.1 | 4 - 5 | 1293 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |