Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bZIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121588.3 | NP_197087.1 | 1 | 996 | 118 | |
GenBank mRNA | AK117828.1 | - | 1 | 665 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |