Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLN1;4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121663.3 | NP_568335.1 | 9 - 10 | 947 | 98 | |
GenBank mRNA | AY059932.1 | - | 9 - 10 | 764 | 98 | |
AY128749.1 | - | 9 - 10 | 721 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |