Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIC40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121668.4 | NP_197165.1 | 2 - 3 | 503 | 63 | |
GenBank mRNA | AF428299.1 | - | 2 - 3 | 425 | 63 | |
AY093010.1 | - | 2 - 3 | 386 | 63 | ||
BT006595.1 | - | 2 - 3 | 315 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |