Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036817.3 | NP_001031894.1 | 4 - 5 | 794 | 142 | |
NM_001343458.1 | NP_001332636.1 | 4 - 5 | 794 | 142 | ||
NM_121669.3 | NP_197166.2 | 4 - 5 | 669 | 142 | ||
GenBank mRNA | AY062755.1 | - | 4 - 5 | 642 | 142 | |
BT010359.1 | - | 3 - 4 | 499 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |