Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHAR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121676.4 | NP_568336.1 | 5 - 6 | 738 | 74 | |
GenBank mRNA | AK118603.1 | - | 5 - 6 | 709 | 74 | |
AK175537.1 | - | 5 - 6 | 710 | 74 | ||
AK175798.1 | - | 5 - 6 | 709 | 74 | ||
AK176039.1 | - | 5 - 6 | 710 | 74 | ||
AK176071.1 | - | 5 - 6 | 699 | 74 | ||
AK176708.1 | - | 5 - 6 | 710 | 74 | ||
AK176870.1 | - | 5 - 6 | 709 | 74 | ||
AY085616.1 | - | 5 - 6 | 710 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |