Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WOX12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203398.1 | NP_001190327.1 | 2 - 3 | 849 | 129 | |
NM_121787.3 | NP_197283.2 | 1 - 2 | 764 | 129 | ||
GenBank mRNA | AK117910.1 | - | 1 - 2 | 686 | 129 | |
AY251403.1 | - | 1 - 2 | 613 | 129 | ||
BT006232.1 | - | 1 - 2 | 613 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |