Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121941.2 | NP_197437.1 | 1 - 2 | 986 | 72 | |
GenBank mRNA | DQ446970.1 | - | 1 - 2 | 812 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |