Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROPGEF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343611.1 | NP_001318601.1 | 2 - 3 | 317 | 69 | |
NM_001343612.1 | NP_001331396.1 | 1 - 2 | 512 | 69 | ||
NM_001343613.1 | NP_001331395.1 | 1 - 2 | 512 | 69 | ||
NM_001343614.1 | NP_001331397.1 | 1 - 2 | 512 | 69 | ||
NM_121961.2 | NP_197457.1 | 1 - 2 | 131 | 69 | ||
GenBank mRNA | DQ459196.1 | - | 1 - 2 | 131 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Arabidopsis thaliana Probe spans exons |